Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.6 mol/L in toluene ≥70%

Supplier: Decon Laboratories Ltd

S0467-25G S0467-500G
TCS0467-25GEA 44 USD InStock
TCS0467-25G TCS0467-500G
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.6 mol/L in toluene ≥70%
Sodium bis(2-methoxyethoxy)aluminium hydride
CAS Number: 22722-98-1
MDL Number: MFCD00011631
Molecular Formula: C6H16AlNaO4
Molecular Weight: 202.16
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 4
Specific Gravity (20/20): 1.02

Other Product Names / Synonyms: Sodium Dihydridobis(2-methoxyethoxy)aluminate


Formula: C₆H₁₆AlNaO₄
Density: 1.02 (20 °C)
Flash Pt: 4 °C
MDL Number: MFCD00011631
CAS Number: 22722-98-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6381d37a568618d4ef25848cffbc5c92d04419a&vid=a6381d37a568618d4ef25848cffbc5c92d04419a&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty