Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Trichloro-2-nitrobenzene ≥96.0%

Supplier: Decon Laboratories Ltd

T0388-25G T0388-5G
TCT0388-25GEA 80.39 USD InStock
TCT0388-25G TCT0388-5G
1,3,5-Trichloro-2-nitrobenzene ≥96.0%
1,3,5-Trichloro-2-nitrobenzene
CAS Number: 18708-70-8
MDL Number: MFCD00014690
Molecular Formula: C6H2Cl3NO2
Molecular Weight: 226.44
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 72


Formula: C₆H₂Cl₃NO₂
Melting Pt: 72 °C
MDL Number: MFCD00014690
CAS Number: 18708-70-8
UN: 2811
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73e9240258b9d271035fbd9b3aceda31102102a7&vid=73e9240258b9d271035fbd9b3aceda31102102a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty