Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,4-Trimethyl-1-pentene ≥98.0%

Supplier: Decon Laboratories Ltd

T0665-100ML T0665-25ML
TCT0665-100MLEA 150.29 USD InStock
TCT0665-100ML TCT0665-25ML
2,4,4-Trimethyl-1-pentene ≥98.0%
2,4,4-Trimethyl-1-pentene
CAS Number: 107-39-1
MDL Number: MFCD00008855
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 100
Melting point (°C): -93
Flash Point (°C): -7
Specific Gravity (20/20): 0.72

Other Product Names / Synonyms: Isooctene


Formula: C₈H₁₆
MW: 112.22
Boiling Pt: 102
Melting Pt: -93.48
Density: 0.72 (20 °C)
Flash Pt: –7 °C
MDL Number: MFCD00008855
CAS Number: 107-39-1
UN: 2050
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b9ac1a089741a984cd5ff67f2a9f156dd8b5838&vid=3b9ac1a089741a984cd5ff67f2a9f156dd8b5838&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty