Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trifluoromethanesulfonyl chloride ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

T1027-25G T1027-5G
TCT1027-25GEA 207.51 USD InStock
TCT1027-25G TCT1027-5G
Trifluoromethanesulfonyl chloride ≥98.0% (by GC)
Trifluoromethanesulphonyl chloride
CAS Number: 421-83-0
MDL Number: MFCD00007451
Molecular Formula: CClF3O2S
Molecular Weight: 168.51
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 31
Specific Gravity (20/20): 1.60
Storage Temperature: 0-10°C


Formula: CClF₃O₂S
Boiling Pt: 31 °C
Density: 1.60 (20 °C)
MDL Number: MFCD00007451
CAS Number: 421-83-0
UN: 3265
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61a4804d5cdbd3fb44e1198f62f800c9818f1c42&vid=61a4804d5cdbd3fb44e1198f62f800c9818f1c42&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty