Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4,5-Tetrafluorobenzene ≥99.0%

Supplier: Decon Laboratories Ltd

T1029-25G T1029-5G
TCT1029-25GEA 49.2 USD InStock
TCT1029-25G TCT1029-5G
1,2,4,5-Tetrafluorobenzene ≥99.0%
1,2,4,5-Tetrafluorobenzene
CAS Number: 327-54-8
MDL Number: MFCD00000307
Molecular Formula: C6H2F4
Molecular Weight: 150.08
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 90
Flash Point (°C): 16
Specific Gravity (20/20): 1.43


Formula: C₆H₂F₄
Boiling Pt: 90 °C
Density: 1.43 (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00000307
CAS Number: 327-54-8
UN: 2387
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c075c16c44837d76a5a65eb793c694b19ddd4e2c&vid=c075c16c44837d76a5a65eb793c694b19ddd4e2c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty