Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trimethylthiazole ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

T1068-25G T1068-5G
TCT1068-25GEA 93.1 USD InStock
TCT1068-25G TCT1068-5G
2,4,5-Trimethylthiazole ≥97.0% (by GC, titration analysis)
2,4,5-Trimethylthiazole
CAS Number: 13623-11-5 MDL Number: MFCD00005332 Molecular Formula: C6H9NS Molecular Weight: 127.21 Purity/Analysis Method: 97.0% (GC,T) Form: Clear Liquid Color: Slightly Pale Yellow Boiling point (°C): 167 Flash Point (°C): 56 Specific Gravity (20/20): 1.04


Formula: C₆H₉NS
Boiling Pt: 167 °C
Density: 1.04 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00005332
CAS Number: 13623-11-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=040ea2f0499a4b1e7016484273262a7daf186cdc&vid=040ea2f0499a4b1e7016484273262a7daf186cdc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty