T1216-25G
T1216-5G
TCT1216-25GEA
1061
USD
InStock
TCT1216-25G
TCT1216-5G
Vinyl trifluoroacetate ≥97.0% stabilised
Vinyl trifluoroacetate
(stabilized with TBC)
CAS Number: 433-28-3
MDL Number: MFCD00003561
Molecular Formula: C4H3F3O2
Molecular Weight: 140.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 42
Flash Point (°C): -31
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Trifluoroacetic Acid Vinyl Ester