Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trimethylsilyl)ethanol ≥95.0%

Supplier: Decon Laboratories Ltd

T1441-25ML T1441-5ML
TCT1441-25MLEA 207.1 USD InStock
TCT1441-25ML TCT1441-5ML
2-(Trimethylsilyl)ethanol ≥95.0%
2-(Trimethylsilyl)ethanol
CAS Number: 2916-68-9
MDL Number: MFCD00002825
Molecular Formula: C5H14OSi
Molecular Weight: 118.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 75
Flash Point (°C): 50
Specific Gravity (20/20): 0.83

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (2-Hydroxyethyl)trimethylsilane


Formula: HOH₂CCH₂Si(CH₃)₃
Boiling Pt: 75 °C
Density: 0.83 (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00002825
CAS Number: 2916-68-9
UN: 1987
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d326c638611a279dadfcf78b841b94ae0de0b47&vid=8d326c638611a279dadfcf78b841b94ae0de0b47&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty