Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyloxonium tetrafluoroborate ≥95.0%

Supplier: Decon Laboratories Ltd

T1507-25G T1507-5G
TCT1507-25GEA 112.47 USD InStock
TCT1507-25G TCT1507-5G
Trimethyloxonium tetrafluoroborate ≥95.0%
Trimethyloxonium tetrafluoroborate
CAS Number: 420-37-1
MDL Number: MFCD00011798
Molecular Formula: C3H9BF4O
Molecular Weight: 147.91
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Storage Temperature: <0°C


Formula: C₃H₉BF₄O
MW: 147.91 g/mol
MDL Number: MFCD00011798
CAS Number: 420-37-1
UN: 3261
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=094d8517ec898d8978dac4b96a9b6f517e913a7b&vid=094d8517ec898d8978dac4b96a9b6f517e913a7b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty