Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)benzylamine ≥98.0%

Supplier: Decon Laboratories Ltd

T1508-25G T1508-5G
TCT1508-25GEA 178 USD InStock
TCT1508-25G TCT1508-5G
4-(Trifluoromethyl)benzylamine ≥98.0%
4-(Trifluoromethyl)benzylamine
CAS Number: 3300-51-4
MDL Number: MFCD00010220
Molecular Formula: C8H8F3N
Molecular Weight: 175.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 113
Flash Point (°C): 75
Specific Gravity (20/20): 1.24

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Aminomethylbenzotrifluoride, alpha-Amino-alpha',alpha',alpha'-trifluoro-p-xylene


Formula: C₈H₈F₃N
Boiling Pt: 113 °C
Density: 1.24 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00010220
CAS Number: 3300-51-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b62a53cd20aa338ac5d8c181948e9fabb1cbe97&vid=4b62a53cd20aa338ac5d8c181948e9fabb1cbe97&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty