Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Trimethylsiloxy)cyclohexene ≥95.0%

Supplier: Decon Laboratories Ltd

T1556-10ML T1556-25ML
TCT1556-10MLEA 87 USD InStock
TCT1556-10ML TCT1556-25ML
1-(Trimethylsiloxy)cyclohexene ≥95.0%
1-(Trimethylsiloxy)cyclohexene
CAS Number: 6651-36-1
MDL Number: MFCD00001541
Molecular Formula: C9H18OSi
Molecular Weight: 170.33
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Flash Point (°C): 41
Specific Gravity (20/20): 0.89

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (1-Cyclohexen-1-yloxy)trimethylsilane


Formula: C₉H₁₈OSi
Boiling Pt: 165 °C
Density: 0.89 (20 °C)
MDL Number: MFCD00001541
CAS Number: 6651-36-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce8e7d1141a44d0a53ddb8f07d31173211bf4a1e&vid=ce8e7d1141a44d0a53ddb8f07d31173211bf4a1e&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty