Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-Trifluorophenol ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

T1616-1G T1616-5G
TCT1616-1GEA 66 USD InStock
TCT1616-1G TCT1616-5G
2,3,4-Trifluorophenol ≥98.0% (by GC)
2,3,4-Trifluorophenol
CAS Number: 2822-41-5
MDL Number: MFCD00042426
Molecular Formula: C6H3F3O
Molecular Weight: 148.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 58
Freezing point (°C): 29


Formula: C₆H₃F₃O
Flash Pt: 58 °C
MDL Number: MFCD00042426
CAS Number: 2822-41-5
UN: 2921
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ec6d8e1a1da945ec43500f701baae6233119be8&vid=5ec6d8e1a1da945ec43500f701baae6233119be8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty