Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,3-Trifluoropropionic acid ≥98.0%

Supplier: Decon Laboratories Ltd

T1713-1G T1713-5G
TCT1713-1GEA 28 USD InStock
TCT1713-1G TCT1713-5G
3,3,3-Trifluoropropionic acid ≥98.0%
3,3,3-Trifluoropropionic acid
CAS Number: 2516-99-6
MDL Number: MFCD00153292
Molecular Formula: C3H3F3O2
Molecular Weight: 128.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 153
Specific Gravity (20/20): 1.47


Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 153 °C
Melting Pt: 0
Density: 1.47 (20 °C)
MDL Number: MFCD00153292
CAS Number: 2516-99-6
UN: 3265
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0565c06f411ee311207e2d1682a78b46d0d70cd1&vid=0565c06f411ee311207e2d1682a78b46d0d70cd1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty