Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α'-Bromo-α,α,α-trifluoro-m-xylene ≥98.0%

Supplier: Decon Laboratories Ltd

T1782-25G T1782-5G
TCT1782-25GEA 139 USD InStock
TCT1782-25G TCT1782-5G
α'-Bromo-α,α,α-trifluoro-m-xylene ≥98.0%
α'-Bromo-α,α,α-trifluoro-m-xylene
CAS Number: 402-23-3
MDL Number: MFCD00000395
Molecular Formula: C8H6BrF3
Molecular Weight: 239.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 79
Flash Point (°C): 88
Specific Gravity (20/20): 1.58

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Bromomethyl)benzotrifluoride, alpha-Bromo-alpha',alpha',alpha'-trifluoro-m-xylene


Formula: C₈H₆BrF₃
MW: 239.03
Boiling Pt: 50
Density: 1.58 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00000395
CAS Number: 402-23-3
UN: 3265
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0ab6ef04345750048c43f40d51fc5139b48979&vid=8e0ab6ef04345750048c43f40d51fc5139b48979&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty