Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o,α,α,α-Tetrafluoroanisole ≥98.0%

Supplier: Decon Laboratories Ltd

T2150-25G T2150-5G
TCT2150-25GEA 89 USD InStock
TCT2150-25G TCT2150-5G
o,α,α,α-Tetrafluoroanisole ≥98.0%
o,α,α,α-Tetrafluoroanisole
CAS Number: 2106-18-5
MDL Number: MFCD00236322
Molecular Formula: C7H4F4O
Molecular Weight: 180.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 105
Specific Gravity (20/20): 1.33


Formula: C₇H₄F₄O
Boiling Pt: 105 °C
Density: 1.33 (20 °C)
MDL Number: MFCD00236322
CAS Number: 2106-18-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06ca4618c8398d7dd2b97db1255a6a0ec1c94500&vid=06ca4618c8398d7dd2b97db1255a6a0ec1c94500&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty