Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethoxy)phenol ≥96.0%

Supplier: Decon Laboratories Ltd

T2156-25G T2156-5G
TCT2156-25GEA 85 USD InStock
TCT2156-25G TCT2156-5G
2-(Trifluoromethoxy)phenol ≥96.0%
2-(Trifluoromethoxy)phenol
CAS Number: 32858-93-8
MDL Number: MFCD00236324
Molecular Formula: C7H5F3O2
Molecular Weight: 178.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.37

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Hydroxyphenyl Trifluoromethyl Ether


Formula: C₇H₅F₃O₂
Density: 1.37 (20 °C)
MDL Number: MFCD00236324
CAS Number: 32858-93-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9e905a3df7dc3793c9e8f542147e9e28fcdb702&vid=b9e905a3df7dc3793c9e8f542147e9e28fcdb702&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty