Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzoyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

T2341-1G T2341-5G
TCT2341-1GEA 29 USD InStock
TCT2341-1G TCT2341-5G
4-(Trifluoromethoxy)benzoyl chloride ≥98.0%
4-(Trifluoromethoxy)benzoyl chloride
CAS Number: 36823-88-8
MDL Number: MFCD00052329
Molecular Formula: C8H4ClF3O2
Molecular Weight: 224.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 56
Flash Point (°C): 90
Specific Gravity (20/20): 1.43


Formula: C₈H₄ClF₃O₂
Boiling Pt: 56 °C
Density: 1.43 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00052329
CAS Number: 36823-88-8
UN: 3265
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1774985b43128628e93cca43592a9b6725862200&vid=1774985b43128628e93cca43592a9b6725862200&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty