Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trimethylpyrylium tetrafluoroborate ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

T2375-25G T2375-5G
TCT2375-25GEA 264 USD InStock
TCT2375-25G TCT2375-5G
2,4,6-Trimethylpyrylium tetrafluoroborate ≥98.0% (by titrimetric analysis)
2,4,6-Trimethylpyrylium tetrafluoroborate
CAS Number: 773-01-3
MDL Number: MFCD00032098
Molecular Formula: C8H11BF4O
Molecular Weight: 209.98
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 208


Formula: C₈H₁₁BF₄O
Melting Pt: 208 °C
MDL Number: MFCD00032098
CAS Number: 773-01-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c785959cdcf3cf8ae86df539c999a80474059ec&vid=9c785959cdcf3cf8ae86df539c999a80474059ec&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty