Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3,4-Tetrafluorobenzene ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

T2410-25G T2410-5G
TCT2410-25GEA 216 USD InStock
TCT2410-25G TCT2410-5G
1,2,3,4-Tetrafluorobenzene ≥98.0% (by GC)
1,2,3,4-Tetrafluorobenzene
CAS Number: 551-62-2
MDL Number: MFCD00000285
Molecular Formula: C6H2F4
Molecular Weight: 150.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 94
Melting point (°C): -42
Flash Point (°C): 20
Specific Gravity (20/20): 1.43


Formula: C₆H₂F₄
Boiling Pt: 94 °C
Melting Pt: –42 °C
Density: 1.43 (20 °C)
Flash Pt: 20 °C
MDL Number: MFCD00000285
CAS Number: 551-62-2
UN: 1993
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0211b813fce1365cdcda8017450c24ca5bd0027&vid=d0211b813fce1365cdcda8017450c24ca5bd0027&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty