T2764-1G
T2764-5G
TCT2764-1GEA
64.08
USD
InStock
TCT2764-1G
TCT2764-5G
Triethyl(trifluoromethyl)silane ≥98.0%
Triethyl(trifluoromethyl)silane
CAS Number: 120120-26-5
MDL Number: MFCD00192520
Molecular Formula: C7H15F3Si
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 58
Flash Point (°C): 0
Specific Gravity (20/20): 0.98
Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: (Triethylsilyl)trifluoromethane