Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)thiobenzamide ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

T2771-25G T2771-5G
TCT2771-25GEA 159 USD InStock
TCT2771-25G TCT2771-5G
4-(Trifluoromethyl)thiobenzamide ≥98.0% (by HPLC)
4-(Trifluoromethyl)thiobenzamide
CAS Number: 72505-21-6
MDL Number: MFCD00051806
Molecular Formula: C8H6F3NS
Molecular Weight: 205.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 137


Formula: C₈H₆F₃NS
Melting Pt: 137 °C
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef23beec46a5794838a0a624ac3a22a331dc4031&vid=ef23beec46a5794838a0a624ac3a22a331dc4031&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty