Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc bromide ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

Z0013-25G Z0013-300G
TCZ0013-25GEA 52 USD InStock
TCZ0013-25G TCZ0013-300G
Zinc bromide ≥98.0% (by titrimetric analysis)
Zinc bromide
CAS Number: 7699-45-8
MDL Number: MFCD00011294
Molecular Formula: Br2Zn
Molecular Weight: 225.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 697
Melting point (°C): 394


Formula: Br₂Zn
Boiling Pt: 697 °C
Melting Pt: 394 °C
MDL Number: MFCD00011294
CAS Number: 7699-45-8
UN: 3260
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf82e53e0b33c6e34dae9354da3ffea74cbb1287&vid=cf82e53e0b33c6e34dae9354da3ffea74cbb1287&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty