Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4,5-Tetrafluorobenzoyl chloride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

121480-1G 121480-25G 121480-5G
BT121480-1GEA 40 USD InStock
BT121480-1G BT121480-25G BT121480-5G
2,3,4,5-Tetrafluorobenzoyl chloride ≥98%
2,3,4,5-Tetrafluorobenzoyl chloride
CAS: 94695-48-4; EC No: 000-000-0; MDL No: MFCD00075164 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C7HClF4O ; MW: 212.53 Flash point: 89°C (192°F) Density (g/mL): 1.580 ; Refractive Index: 1.479 Moisture Sensitive

  • Refractive index: 1.479
  • Moisture Sensitive
  • Liquid


Formula: C₇HClF₄O
Density: 1.580 g/mL
Flash Pt: 89 °C (192 °F)
MDL Number: MFCD00075164
CAS Number: 94695-48-4
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty