Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mercury(II) nitrate monohydrate ≥98.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

121540-250G 121540-50G
BT121540-250GEA 242.65 USD InStock
BT121540-250G BT121540-50G
Mercury(II) nitrate monohydrate ≥98.0% ACS
Mercury(II) nitrate monohydrate
CAS: 7783-34-8; EC No: 233-886-4; MDL No: MFCD00149736 UN No: UN1625; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Hg(NO3)2·xH2O; MW: 324.60 (anhydrous) Melting Point: 79° Density (g/mL): 4.39 Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: Hg(NO₃)₂·1H₂O
MW: 342.62 g/mol
Melting Pt: 79
Density: 4.39 g/mL
MDL Number: MFCD00149736
CAS Number: 7783-34-8
EINECS: 233-886-4
UN: 1625
ADR: 6.1 II
Order Now
Specification Test Results
Assay 98.0% min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty