Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

121555-100G 121555-25G 121555-5G
BT121555-100GEA 495 USD InStock
BT121555-100G BT121555-25G BT121555-5G
N-Methyl-N-(trimethylsilyl)trifluoroacetamide ≥97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
CAS: 24589-78-4; EC No: 246-331-6; MDL No: MFCD00000411; RTECS: AC9800000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CF3CON(CH3)Si(CH3)3; Molecular Formula: C6H12F3NOSi; MW: 199.25 Boiling Point: 130-132°; Flash point: 25°C (77°F) Density (g/mL): 1.075; Refractive Index: 1.380 Moisture Sensitive

  • RTECS: AC9800000
  • Refractive index: 1.380
  • Moisture Sensitive
  • Liquid


Formula: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Boiling Pt: 130
Density: 1.075 g/mL
Flash Pt: 25 °C (77 °F)
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6
ADR: 3 III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty