Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethyldisilane ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

122625-100G 122625-25G 122625-5G
BT122625-100GEA 291 USD InStock
BT122625-100G BT122625-25G BT122625-5G
Hexamethyldisilane ≥98%
Hexamethyldisilane
CAS: 1450-14-2; EC No: 215-911-0; MDL No: MFCD00008258; RTECS: JM9170000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3SiSi(CH3)3; Molecular Formula: C6H18Si2; MW: 146.38 Melting Point: 9-12°; Boiling Point: 112-114°; Flash point: 11°C (52°F) Density (g/mL): 0.715; Refractive Index: 1.422

  • RTECS: JM9170000
  • Refractive index: 1.422
  • Liquid


Formula: C₆H₁₈Si₂
MW: 146.38 g/mol
Boiling Pt: 112…114 °C
Melting Pt: 9…12 °C
Density: 0.715 g/mL
Flash Pt: 11 °C (52 °F)
MDL Number: MFCD00008258
CAS Number: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty