Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrapropylammonium perruthenate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

122915-1G 122915-250MG
BT122915-1GEA 142.91 USD InStock
BT122915-1G BT122915-250MG
Tetrapropylammonium perruthenate ≥98%
Tetrapropylammonium perruthenate
CAS: 114615-82-6; MDL No: MFCD00074914 UN No: UN1479; Haz Class: 5.1; Packing Group: III Solid; Linear Formula: (CH3CH2CH2)4N(RuO4); Molecular Formula: C12H28NO4Ru ; MW: 351.43 Melting Point: 160° (decomposes) Hygroscopic

  • Hygroscopic
  • Solid


Formula: C₁₂H₂₈NO₄Ru
Melting Pt: 160 °C (dec.)
MDL Number: MFCD00074914
CAS Number: 114615-82-6
UN: 1479
ADR: 5.1,III
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty