Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-chlorobutane ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

123005-100G 123005-25G
BT123005-100GEA 140 USD InStock
BT123005-100G BT123005-25G
1-Bromo-4-chlorobutane ≥99%
1-Bromo-4-chlorobutane
CAS: 6940-78-9; EC No: 230-089-3; MDL No: MFCD00001010 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: Br(CH2)4Cl; MW: 171.46 Boiling Point: 80-82°/30 mmHg; Flash point: 60°C (140°F) Density (g/mL): 1.488; Refractive Index: 1.488

  • Refractive index: 1.488
  • Liquid


Formula: BrH₂C(CH₂)₂CH₂Cl
MW: 171.46 g/mol
Boiling Pt: 80
Density: 1.488 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00001010
CAS Number: 6940-78-9
EINECS: 230-089-3
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty