Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis(acetonitrile)palladium(II) tetrafluoroborate

Supplier: BEANTOWN CHEMICAL - BTC

123030-1G 123030-250MG
BT123030-1GEA 110 USD InStock
BT123030-1G BT123030-250MG
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
CAS: 21797-13-7; MDL No: MFCD00043297 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: Pd(CH3CN)4(BF4)2; Molecular Formula: C8H12B2F8N4Pd; MW: 444.22 Melting Point: 230° (decomposes) Air Sensitive, Moisture Sensitive

  • Air Sensitive, Moisture Sensitive
  • Powder


Formula: C₈H₁₂B₂F₈N₄Pd
MW: 444.239 g/mol
Melting Pt: 230 °C (dec.)
MDL Number: MFCD00043297
CAS Number: 21797-13-7
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty