123065-100G
123065-25G
123065-5G
BT123065-100GEA
354.84
USD
InStock
BT123065-100G
BT123065-25G
BT123065-5G
Bismuth ≥99.999% (trace metals basis), powder -200 mesh
Bismuth
CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033; RTECS: EB2600000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Bi; MW: 208.98
Melting Point: 271°; Boiling Point: 1560°
Density (g/mL): 9.8