Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobornyl acrylate ≥85% stabilised, Technical Grade

Supplier: BEANTOWN CHEMICAL - BTC

123080-250G 123080-50G
BT123080-250GEA 55 USD InStock
BT123080-250G BT123080-50G
Isobornyl acrylate ≥85% stabilised, Technical Grade
Isobornyl acrylate
CAS: 5888-33-5; EC No: 227-561-6; MDL No: MFCD00080424; RTECS: UD3940000 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C13H20O2; MW: 208.3 Boiling Point: 119-121°/15 mmHg; Flash point: 108°C (226°F) Density (g/mL): 0.986; Refractive Index: 1.476 Light Sensitive

  • RTECS: UD3940000
  • Refractive index: 1.476
  • Light Sensitive
  • Liquid


Formula: C₁₃H₂₀O₂
MW: 208.30 g/mol
Boiling Pt: 119…121 °C/15 mmHg
Density: 0.986 g/mL
Flash Pt: 108 °C (226 °F)
MDL Number: MFCD00080424
CAS Number: 5888-33-5
EINECS: 227-561-6
ADR: 9 III
Order Now
Specification Test Results
Assay 85%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty