Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyltrimethylsilane ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

123875-10G 123875-250G 123875-50G
BT123875-10GEA 45.2 USD InStock
BT123875-10G BT123875-250G BT123875-50G
Allyltrimethylsilane ≥98%
Allyltrimethylsilane


Formula: H₂CCHCH₂Si(CH₃)₃
Boiling Pt: 84…88 °C
Density: 0.719 g/mL
MDL Number: MFCD00008635
CAS Number: 762-72-1
EINECS: 212-104-5
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty