123905-10G
123905-50G
BT123905-10GEA
67.35
USD
InStock
BT123905-10G
BT123905-50G
Sodium deuteroxide 40% (w/w) in D₂O , 99.5% (Isotopic)
Sodium deuteroxide
CAS: 14014-06-3; EC No: 237-825-2; MDL No: MFCD00037669
UN No: UN1824; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: NaOD; MW: 41.00
Density (g/mL): 1.516; Refractive Index: 1.418
Air Sensitive, Hygroscopic, Moisture Sensitive
- Refractive index: 1.418
- Air Sensitive, Hygroscopic, Moisture Sensitive
- Liquid