Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Bromopropionic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

124145-1KG 124145-250G
BT124145-1KGEA 148 USD InStock
BT124145-1KG BT124145-250G
(±)-2-Bromopropionic acid ≥98%
(±)-2-Bromopropionic acid
CAS: 598-72-1; EC No: 209-947-6; MDL No: MFCD00004211; RTECS: UA2451715 UN No: UN3261; Haz Class: 8; Packing Group: II Liquid/Low Melting Solid; Linear Formula: CH3CHBrCO2H; Molecular Formula: C3H5BrO2; MW: 152.98 Boiling Point: 203°; Flash point: 100°C (212°F) Density (g/mL): 1.7; Refractive Index: 1.475

  • RTECS: UA2451715
  • Refractive index: 1.475
  • Liquid/Low Melting Solid


Formula: H₃CBrCHCOOH
MW: 152.98 g/mol
Boiling Pt: 203
Melting Pt: 25
Density: 1.7 g/mL
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00004211
CAS Number: 598-72-1
EINECS: 209-947-6
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty