Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diantimony triselenide ≥99.999%

Supplier: BEANTOWN CHEMICAL - BTC

124245-25G 124245-5G
BT124245-25GEA 438 USD InStock
BT124245-25G BT124245-5G
Diantimony triselenide ≥99.999%
Diantimony triselenide
CAS: 1315-05-5; EC No: 215-256-0; MDL No: MFCD00016319 UN No: UN1549; Haz Class: 6.1; Packing Group: III 3-12mm Pieces; Linear Formula: Sb2Se3; MW: 480.40 Moisture Sensitive

  • Moisture Sensitive
  • 3-12mm Pieces


Formula: Sb₂Se₃
MW: 480.4
Melting Pt: 605
Density: 5.81
MDL Number: MFCD00016319
CAS Number: 1315-05-5
EINECS: 215-256-0
UN: 1549
ADR: 6.1 III
Merck Index: 12,00753
Order Now
Specification Test Results
Assay 99.999%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty