Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc chloride ≥97.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

124275-1KG 124275-250G
BT124275-1KGEA 173.54 USD InStock
BT124275-1KG BT124275-250G
Zinc chloride ≥97.0% ACS
Zinc chloride
CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295; RTECS: ZH1400000 UN No: UN2331; Haz Class: 8; Packing Group: III Powder; Linear Formula: ZnCl2; MW: 136.29 Melting Point: 293°; Boiling Point: 732° Density (g/mL): 2.907 Hygroscopic

  • RTECS: ZH1400000
  • Hygroscopic
  • Powder


Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.907 g/mL
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185
Order Now
Specification Test Results
Assay 97.0% min.
Oxycloride Passes test
Insoluble matter ≤0.005%
Nitrate (NO3) ≤0.003%
Sulfate (SO4) ≤0.01%
Ammonium (NH4) ≤0.005%
Calcium (Ca) ≤0.06%
Iron (Fe) ≤0.001%
Lead (Pb) ≤0.005%
Magnesium (Mg) ≤0.01%
Potassium (K) ≤0.02%
Sodium (Na) ≤0.05%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty