Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylphosphonium hydroxide 40% (w/w) in water

Supplier: BEANTOWN CHEMICAL - BTC

124460-1L 124460-250ML 124460-50ML
BT124460-1LEA 715 USD InStock
BT124460-1L BT124460-250ML BT124460-50ML
Tetrabutylphosphonium hydroxide 40% (w/w) in water
Tetrabutylphosphonium hydroxide
CAS: 14518-69-5; EC No: 238-528-0; MDL No: MFCD00068456 UN No: UN3267; Haz Class: 8; Packing Group: II Liquid; Linear Formula: [CH3(CH2)3]4P(OH); Molecular Formula: C16H37OP; MW: 276.45 Density (g/mL): 0.989; Refractive Index: 1.412

  • Refractive index: 1.412
  • Liquid


Formula: C₁₆H₃₇OP
MW: 276.45 g/mol
Density: 0.989 g/mL
MDL Number: MFCD00068456
CAS Number: 14518-69-5
EINECS: 238-528-0
UN: 3267
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty