Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium ≥97.5%, spherical powder APS 3.0-4.5 µm

Supplier: BEANTOWN CHEMICAL - BTC

124525-1KG 124525-250G 124525-50G
BT124525-1KGEA 136 USD InStock
BT124525-1KG BT124525-250G BT124525-50G
Aluminium ≥97.5%, spherical powder APS 3.0-4.5 µm
Aluminium
CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 UN No: UN1396; Haz Class: 4.3; Packing Group: II Powder; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7

  • RTECS: BD0330000
  • Powder


Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3 II
Merck Index: 13,00321
Order Now
Specification Test Results
Assay 97.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty