Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diamminepalladium(II) nitrite (8 - 10% Pd) (w/w) in water

Supplier: BEANTOWN CHEMICAL - BTC

124550-C2G 124550-C500MG
BT124550-C2GEA 406 USD InStock
BT124550-C2G BT124550-C500
Diamminepalladium(II) nitrite (8 - 10% Pd) (w/w) in water
Diamminepalladium(II) nitrite
CAS: 28068-05-5; MDL No: MFCD00038712 UN No: UN2672; Haz Class: 8; Packing Group: III Liquid; Linear Formula: (NH3)2Pd(NO2)2; Molecular Formula: H6N4O4Pd; MW: 232.47 (anhydrous)

  • Liquid


Formula: (NH₃)₂Pd(NO₂)₂
MW: 232.47 g/mol
MDL Number: MFCD00038712
CAS Number: 28068-05-5
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty