Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenylsilane ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

124565-100G 124565-25G 124565-5G
BT124565-100GEA 981.08 USD InStock
BT124565-100G BT124565-25G BT124565-5G
Phenylsilane ≥97%
Phenylsilane
CAS: 694-53-1; EC No: 211-772-5; MDL No: MFCD00013478 UN No: UN3399; Haz Class: 4.3 (3); Packing Group: II Liquid; Linear Formula: C6H5SiH3; MW: 108.22 Boiling Point: 120°; Flash point: 7°C (45°F) Density (g/mL): 0.877; Refractive Index: 1.510 Moisture Sensitive

  • Refractive index: 1.510
  • Moisture Sensitive
  • Liquid


Formula: C₆H₈Si
MW: 108.22 g/mol
Boiling Pt: 118
Density: 0.877 g/mL
Flash Pt: 7 °C (45 °F)
MDL Number: MFCD00013478
CAS Number: 694-53-1
EINECS: 211-772-5
UN: 3399
ADR: 4.3 II
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty