Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethylamine ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

124830-100ML 124830-2.5L 124830-500ML
BT124830-100MLEA 26.21 USD InStock
BT124830-100ML BT124830-2.5L BT124830-500ML
Diethylamine ≥99%
Diethylamine
CAS: 109-89-7; EC No: 203-716-3; MDL No: MFCD00009032; RTECS: HZ8750000 UN No: UN1154; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: (CH3CH2)2NH; Molecular Formula: C4H11N; MW: 73.14 Melting Point: -50°; Boiling Point: 55°; Flash point: -23°C (-9°F) Density (g/mL): 0.707; Refractive Index: 1.385 Air Sensitive

  • RTECS: HZ8750000
  • Refractive index: 1.385
  • Air Sensitive
  • Liquid


Formula: (C₂H₅)₂NH
Boiling Pt: 55 °C
Melting Pt: –50 °C
Density: 0.707 g/mL
Flash Pt: –23 °C (–9 °F)
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=3c054fa528239af221505ef08b7c0d2503c9d1d7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty