Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodoethanol ≥99% stabilized

Supplier: BEANTOWN CHEMICAL - BTC

125070-100G 125070-25G 125070-5G
BT125070-100GEA 352 USD InStock
BT125070-100G BT125070-25G BT125070-5G
2-Iodoethanol ≥99% stabilized
2-Iodoethanol
CAS: 624-76-0; EC No: 210-861-6; MDL No: MFCD00002830; RTECS: KL3820000 UN No: UN2810; Haz Class: 6.1; Packing Group: I Liquid; Linear Formula: ICH2CH2OH; Molecular Formula: C2H5IO; MW: 171.97 Boiling Point: 85°/25 mmHg; Flash point: 65°C (149°F) Density (g/mL): 2.205; Refractive Index: 1.572 Light Sensitive

  • RTECS: KL3820000
  • Refractive index: 1.572
  • Light Sensitive
  • Liquid


Formula: C₂H₅IO
Boiling Pt: 85 °C/25 mmHg
Density: 2.205 g/mL
Flash Pt: 65 °C (149 °F)
MDL Number: MFCD00002830
CAS Number: 624-76-0
EINECS: 210-861-6
UN: 2810
ADR: 6.1,I
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty