Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrogen peroxide-urea ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

125840-100G 125840-25G 125840-500G
BT125840-100GEA 52.1 USD InStock
BT125840-100G BT125840-25G BT125840-500G
Hydrogen peroxide-urea ≥97%
Hydrogen peroxide-urea
CAS: 124-43-6; EC No: 204-701-4; MDL No: MFCD00013119 UN No: UN1511; Haz Class: 5.1 (8); Packing Group: III Crystalline; Linear Formula: CO(NH2)2·H2O2 ; Molecular Formula: CH4N2O·H2O2 ; MW: 94.07 Melting Point: 90-93° Density (g/mL): 1.39 Air Sensitive, Light Sensitive, Moisture Sensitive

  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Crystalline


Formula: CH₆N₂O₃
Melting Pt: 90…93 °C
Density: 1.39 g/mL
MDL Number: MFCD00013119
CAS Number: 124-43-6
EINECS: 204-701-4
UN: 1511
ADR: 5.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty