Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper germanium selenide ≥99.99%

Supplier: BEANTOWN CHEMICAL - BTC

126315-10G 126315-50G
BT126315-10GEA 197 USD InStock
BT126315-10G BT126315-50G
Copper germanium selenide ≥99.99%
Copper germanium selenide
MDL No: MFCD12546179 UN No: UN3283; Haz Class: 6.1; Packing Group: III 3-6mm Pieces; Molecular Formula: CuGeSe; MW: 215.12

  • 3-6mm Pieces


Formula: CuGeSe
MDL Number: MFCD12546179
CAS Number: 54723-90-9
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty