Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium 98, powder_spherical APS 10.0-14.0 µm

Supplier: BEANTOWN CHEMICAL - BTC

126690-1KG 126690-250G 126690-5KG
BT126690-1KGEA 103 USD InStock
BT126690-1KG BT126690-250G BT126690-5KG
Aluminium 98, powder_spherical APS 10.0-14.0 µm
Aluminium
CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029; RTECS: BD0330000 UN No: UN1396; Haz Class: 4.3; Packing Group: II Powder; Molecular Formula: Al; MW: 26.98 Melting Point: 660.4°; Boiling Point: 2460° Density (g/mL): 2.7

  • RTECS: BD0330000
  • Powder


Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/mL
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3 II
Merck Index: 13,00321
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty