Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutyl chloroformate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

126750-100G 126750-25G 126750-500G
BT126750-100GEA 37.89 USD InStock
BT126750-100G BT126750-25G BT126750-500G
Isobutyl chloroformate ≥98%
Isobutyl chloroformate
CAS: 543-27-1; EC No: 208-840-1; MDL No: MFCD00000642 UN No: UN2742; Haz Class: 6.1 (8,3); Packing Group: II Liquid; Molecular Formula: C5H9ClO2; MW: 136.58 Boiling Point: 128-129°; Flash point: 30°C (86°F) Density (g/mL): 1.053; Refractive Index: 1.406 Moisture Sensitive

  • Refractive index: 1.406
  • Moisture Sensitive
  • Liquid


Formula: C₅H₉ClO₂
Boiling Pt: 128…129 °C
Density: 1.053 g/mL
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD00000642
CAS Number: 543-27-1
EINECS: 208-840-1
UN: 2742
ADR: 6.1,II
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty