Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vanadium(IV) oxide phthalocyanine

Supplier: BEANTOWN CHEMICAL - BTC

126910-10G 126910-2G 126910-500MG
BT126910-10GEA 278 USD InStock
BT126910-10G BT126910-2G BT126910-500MG
Vanadium(IV) oxide phthalocyanine
Vanadium(IV) oxide phthalocyanine
CAS: 13930-88-6; EC No: 237-700-2; MDL No: MFCD00041966 UN No: UN3285; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C32H16N8OV ; MW: 579.45 Hygroscopic

  • Hygroscopic
  • Crystalline


Formula: C₃₂H₁₆N₈OV
MW: 579.45 g/mol
MDL Number: MFCD00041966
CAS Number: 13930-88-6
EINECS: 237-700-2
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty