Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triclosan ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

127385-100G 127385-25G 127385-500G
BT127385-100GEA 132 USD InStock
BT127385-100G BT127385-25G BT127385-500G
Triclosan ≥99%
Triclosan
CAS: 3380-34-5; EC No: 222-182-2; MDL No: MFCD00800992 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: C12H7Cl3O2; MW: 289.54 Melting Point: 56-58°

  • Powder


Formula: C₁₂H₇Cl₃O₂
MW: 289.54 g/mol
Melting Pt: 56…58 °C
MDL Number: MFCD00800992
CAS Number: 3380-34-5
EINECS: 222-182-2
ADR: 9 III
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty