Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydrochloric acid 50% (v/v) in water

Supplier: BEANTOWN CHEMICAL - BTC

127920-1L 127920-4L 127920-500ML
BT127920-1LEA 61.76 USD InStock
BT127920-1L BT127920-4L BT127920-500ML
Hydrochloric acid 50% (v/v) in water
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000 UN No: UN1789; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: HCl ; MW: 36.46

  • RTECS: MW4025000
  • Liquid


Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty