Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium ≥99%, shot ≤1 cm

Supplier: BEANTOWN CHEMICAL - BTC

128130-100G 128130-500G
BT128130-100GEA 41.55 USD InStock
BT128130-100G BT128130-500G
Calcium ≥99%, shot ≤1 cm
Calcium
CAS: 7440-70-2; EC No: 213-179-5; MDL No: MFCD00085314; RTECS: EV8040000 UN No: UN1401; Haz Class: 4.3; Packing Group: II Solid; Molecular Formula: Ca; MW: 40.08 Melting Point: 850°; Boiling Point: 1484° Density (g/mL): 1.54

  • RTECS: EV8040000
  • Solid


Formula: Ca
MW: 40.08 g/mol
Boiling Pt: 1484
Melting Pt: 850
Density: 1.54 g/mL
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3 II
Merck Index: 13,01644
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty